You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Home
Shop All Products
Protein Analysis Reagents
Crosslinking, Labeling and Protein Modification
Crosslinking Reagents
Sulfo-SIAB (sulfosuccinimidyl (4-iodoacetyl)aminobenzoate)
Thermo Scientific™
Sulfo-SIAB (sulfosuccinimidyl (4-iodoacetyl)aminobenzoate)
Catalog number:
22327
Related applications:
Protein Labeling & Crosslinking
Technical Support
Customer Service
Thermo Scientific™
Sulfo-SIAB (sulfosuccinimidyl (4-iodoacetyl)aminobenzoate)
Catalog number:
22327
Related applications:
Protein Labeling & Crosslinking
Technical Support
Customer Service
Catalog Number
22327
Unit Size
50 mg
Price (CLP)
Contact Us ›
Save to list
Catalog Number
Unit Size
Price (CLP)
22327
50 mg
Contact Us ›
Save to list
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Sulfo-SIAB a water-soluble, mid-length (10.6 angstrom) crosslinker for amine-to-sulfhydryl conjugation via sulfo-N-hydroxysuccinimide (sulfo-NHS) ester and iodoacetyl reactive groups.
Features of Sulfo-SIAB:
•
Reactive groups:
sulfo-NHS ester
and
iodoacetyl
•
Reactive toward:
amino and sulfhydryl groups
• Non-cleavable
• Water-soluble (compare to
SIAB
)
Product References:
Crosslinker Application Guide -- search for recent literature references for this product
Related Products
SIAB (succinimidyl (4-iodoacetyl)aminobenzoate)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Reactivity
Amine-Sulfhydryl
Cleavable
No
Description
Sulfo-SIAB
Molecular Weight (g/mol)
504.19
PEGylated
No
Spacer Arm Length
10.6 Å
Cell Permeability
No
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
Haloacetyl, Sulfo-NHS Ester
Spacer
Medium (10 to 30 Å)
Form
Powder
Quantity
50 mg
Solubility
Water
Format
Standard
Water Soluble
Yes
Product Type
Crosslinker
Contents & Storage
Upon receipt store at -20°C protected from light.
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search