Thermo Scientific™

SIAB (succinimidyl (4-iodoacetyl)aminobenzoate)

Catalog number: 22329
Have Questions?
Thermo Scientific™

SIAB (succinimidyl (4-iodoacetyl)aminobenzoate)

Catalog number: 22329
Have Questions?
Catalog Number
22329
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2232950 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce SIAB is a mid-length (10.6 angstrom) crosslinker for amine-to-sulfhydryl conjugation via N-hydroxysuccinimide (NHS) ester and iodoacetyl reactive groups.

Features of SIAB:

Reactive groups: NHS ester and iodoacetyl
Reactive toward: amino and sulfhydryl groups
• Non-cleavable
• Water-insoluble (dissolve first in DMF or DMSO); compare to Sulfo-SIAB
• Used to prepare stable enzyme-IgG conjugates
• Forms conjugates with liposomes

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
Sulfo-SIAB (sulfosuccinimidyl (4-iodoacetyl)aminobenzoate)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine-Sulfhydryl
Cleavable
No
Description
SIAB
Molecular Weight (g/mol)
402.14
PEGylated
No
Spacer Arm Length
10.6 Å
Cell Permeability
Yes
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
Haloacetyl, NHS Ester
Spacer
Medium (10 to 30 Å)
Form
Powder
Quantity
50 mg
Solubility
DMF, DMSO
Format
Standard
Water Soluble
No
Product Type
Crosslinker

Contents & Storage

Upon receipt store at 4°C protected from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text