Thermo Scientific™

BMB (1,4-bismaleimidobutane)

Catalog number: 22331
Have Questions?
Thermo Scientific™

BMB (1,4-bismaleimidobutane)

Catalog number: 22331
Have Questions?
Catalog Number
22331
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2233150 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce BMB is a mid-length, maleimide crosslinker for covalent, irreversible conjugation between sulfhydryl groups (e.g., protein or peptide cysteines).

Features of BMB:

Reactive groups: maleimide (both ends)
Reactive towards: sulfhydryl groups
• Mid-length (10.9Å), sulfhydryl-to-sulfhydryl crosslinker, composed of maleimide groups and 9-atom spacer arm
• Water-insoluble—dissolve first in DMF or DMSO, then add to aqueous reaction buffers
• Noncleavable

Related Products
BMH (bismaleimidohexane)
BMOE (bismaleimidoethane)
BM(PEG)2 (1,8-bismaleimido-diethyleneglycol)
BM(PEG)3 (1,11-bismaleimido-triethyleneglycol)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Sulfhydryl-Sulfhydryl
Cleavable
No
Description
BMB
Molecular Weight (g/mol)
248.23
PEGylated
No
Spacer Arm Length
10.9 Å
Cell Permeability
Yes
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Homobifunctional
Reactive Moiety
Maleimide
Spacer
Medium (10 to 30 Å)
Form
Solid
Quantity
50 mg
Solubility
DMF, DMSO
Format
Standard
Water Soluble
No
Product Type
Crosslinker

Contents & Storage

Upon receipt store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text