Thermo Scientific Chemicals

Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals

Catalog number: 223312500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Perchloric acid, for analysis, ca. 70% solution in water, Thermo Scientific Chemicals

Catalog number: 223312500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 223312500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7601-90-3
IUPAC Name
perchloric acid
Molecular Formula
ClHO4
InChI Key
VLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILES
O[Cl](=O)(=O)=O
Heavy metals (as Pb)
=<1 ppm
Iron (Fe)
=<1 ppm
Silicate (as SiO2)
=<0.0005 %
Titration with NaOH
69.0 to 72.0 %
Sulfate (SO4)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text