Thermo Scientific™

BM(PEG)3 (1,11-bismaleimido-triethyleneglycol)

Catalog number: 22337
Have Questions?
Thermo Scientific™

BM(PEG)3 (1,11-bismaleimido-triethyleneglycol)

Catalog number: 22337
Have Questions?
Catalog Number
22337
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2233750 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce BM(PEG)3 is a bismaleimide-activated PEG compound for crosslinking between sulfhydryl (—SH) groups in proteins and other thiol molecules. The maleimide groups at either end of the PEG3 spacer react specifically and efficiently with reduced sulfhydryls at pH 6.5-7.5 to form stable thioether bonds.

Features of BM(PEG)3:

Reactive groups: maleimide (both ends)
Reactive towards: sulfhydryl groups
• Long, pegylated, sulfhydryl-to-sulfhydryl crosslinkers, composed of maleimide groups and 3-unit polyethylene glycol spacer arm
• PEG spacers provide unique advantages, including enhanced solubility, increased stability, reduced tendency toward aggregation and reduced immunogenicity
• Pure compounds with defined structure and molecular weight, ensuring reproducible protein-modification effects
• Ideal for small molecule or peptide conjugations

Alternative Names for BM(PEG)n:

• Bismaleimide-dPEG
• Mal-PEG-Mal
• BM(PEO)2
• BM-PEO3

Product References:

Crosslinker Application Guide -- search for recent literature references for this product

Related Products
BMB (1,4-bismaleimidobutane)
BMH (bismaleimidohexane)
BMOE (bismaleimidoethane)
BM(PEG)2 (1,8-bismaleimido-diethyleneglycol)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Sulfhydryl-Sulfhydryl
Cleavable
No
Description
BM(PEG)3
Molecular Weight (g/mol)
352.34
PEGylated
Yes
Spacer Arm Length
17.8 Å
Cell Permeability
No
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Homobifunctional
Reactive Moiety
Maleimide
Spacer
Medium (10 to 30 Å)
Form
Solid
Quantity
50 mg
Solubility
Water
Format
Standard
Water Soluble
Yes
Product Type
Crosslinker

Contents & Storage

Upon receipt store desiccated at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text