Saccharin, sodium salt hydrate, 99+%
Saccharin, sodium salt hydrate, 99+%
Saccharin, sodium salt hydrate, 99+%
Saccharin, sodium salt hydrate, 99+%
Thermo Scientific Chemicals

Saccharin, sodium salt hydrate, 99+%

CAS: 82385-42-0 | C7H4NNaO3S | 205.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 223372500
Price (USD)
60.65
Online Exclusive
67.60
Save 6.95 (10%)
Each
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
60.65
Online Exclusive
67.60
Save 6.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS82385-42-0
IUPAC Namesodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate
Molecular FormulaC7H4NNaO3S
InChI KeyOAZGZMKGRPRHJX-UHFFFAOYSA-M
SMILES[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Benzoate and salicylatePasses test
Benzoate and salicylatePasses test
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0468986Certificate of AnalysisJan 31, 2025223372500, 223370051, 223370010
A0465093Certificate of AnalysisAug 15, 2024223372500, 223370051, 223370010
A0460424Certificate of AnalysisFeb 27, 2024223372500, 223370051, 223370010
A0453431Certificate of AnalysisAug 25, 2023223372500, 223370051, 223370010
A0452723Certificate of AnalysisMay 03, 2023223372500, 223370051, 223370010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection