Thermo Scientific™

SBAP (succinimidyl 3-(bromoacetamido)propionate)

Catalog number: 22339
Have Questions?
Thermo Scientific™

SBAP (succinimidyl 3-(bromoacetamido)propionate)

Catalog number: 22339
Have Questions?
Catalog Number
22339
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2233950 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce SBAP is a short (6.2 angstrom) crosslinker for amine-to-sulfhydryl conjugation via N-hydroxysuccinimide (NHS) ester and bromoacetyl reactive groups.

Features of SBAP:

Reactive groups: NHS ester and bromoacetyl
Reactive toward: amino and sulfhydryl groups
• NHS ester reacts with primary amines at pH 7-9 to form a stable amide bond
• Bromacetyl group reacts with sulfhydryl groups at pH > 7.5 to form stable thioether bonds
• Non-cleavable
• Water-insoluble (dissolve first in DMF or DMSO)
• Spacer maintains peptide-like character in the crosslinked species
• Resulting crosslink is susceptible to acid hydrolysis
• Useful for preparing cyclic peptides and peptide conjugates

Product References:

Crosslinker Application Guide -- search for recent literature references for this product
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine-Sulfhydryl
Cleavable
No
Description
SBAP
Molecular Weight (g/mol)
307.1
PEGylated
No
Spacer Arm Length
6.2 Å
Cell Permeability
Yes
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
Haloacetyl, NHS Ester
Spacer
Short (<10 Å)
Form
Solid
Quantity
50 mg
Solubility
DMF, DMSO
Format
Standard
Water Soluble
No
Product Type
Crosslinker
Unit Size
50 mg

Contents & Storage

Upon receipt store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text