Thermo Scientific™

MS(PEG)4 Methyl-PEG-NHS-Ester Reagent

Catalog number: 22342
Have Questions?
Thermo Scientific™

MS(PEG)4 Methyl-PEG-NHS-Ester Reagent

Catalog number: 22342
Have Questions?
Catalog Number
22342
Unit Size
1 g
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
223421 g
-
22341100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce MS(PEG)n reagents are methyl-terminated, polyethylene glycol compounds (n equals 4 to 24 PEG units) activated as NHS esters for covalent pegylation of primary amines on proteins (e.g., lysines) or assay surfaces.

Features of MS(PEG)4:

• NHS-activated for efficient PEGylation of primary amines at pH 7-9; reaction of NHS-ester group results in formation of stable, irreversible amide bonds
• Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
• Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
• PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
• Easy-to-follow instructions increase the likelihood of a successful outcome

MS(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with methyl (—CH3) and amine-reactive NHS-ester groups at opposite ends. The unbranched, hydrophilic, discrete-length molecules have the form Methyl-PEGn-NHS Ester, where the subscript 'n' denotes 4, 8, 12, or 24 ethylene glycol units. The N-hydroxysuccinimide (NHS) ester is spontaneously reactive with primary amines (—NH2), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.

PEGylation Applications:
• PEGylate amine surfaces
• Add inert mass to proteins, immunogens, drug compounds and probes
• Improve solubility (decrease aggregation) of proteins or peptides without affecting function
• Protect proteins from proteolysis

Why PEGylate a protein or peptide?
PEG-containing reagents have been used to modify proteins to provide specific advantages. Protein PEGylation can improve the stability of the modified protein, protect it from proteolytic digestion, increase its half life in biological applications, mask it from causing an immunogenic response, decrease its antigenicity or potential toxicity, improve its solubility, diminish the potential for aggregation, and minimize interference for both in vitro and in vivo applications. Polyethylene glycol, also called polyethylene oxide (PEO), has these effects because it is nontoxic, nonimmunogenic, hydrophilic, water soluble and highly flexible.

Advantages of Discrete-length mPEG-NHS Ester Compounds
These reagents are specially synthesized as homogeneous compounds of discrete chain length and defined molecular weight. As such, they enable precise control and optimization of surface protein modification experiments. By contrast, typical preparations of PEG compounds are heterogeneous mixtures composed of multiple chain lengths and a range of molecular weights.

Related Products
MS(PEG)8 Methyl-PEG-NHS-Ester Reagent
MS(PEG)12 Methyl-PEG-NHS-Ester Reagent
MS(PEG)24 Methyl-PEG-NHS-Ester Reagent
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine
Description
Pierce MS(PEG)4
Label Type
Polyethylene Glycol (PEG), Blocking Groups
Reactive Moiety
Active Ester, Succinimidyl Ester, NHS Ester
Spacer
4 ethylene glycol units
Form
Solid
Quantity
1 g
Solubility
Water
Product Type
NHS-Ester Reagent

Contents & Storage

Store desiccated at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text