3,3-Dimethyl-1-butyne, 98%, Thermo Scientific Chemicals
3,3-Dimethyl-1-butyne, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3-Dimethyl-1-butyne, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number 223440250
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
Add to cart
3,3-Dimethyl-1-butyne, 98%, Thermo Scientific Chemicals
Catalog number223440250
Price (USD)/ Each
185.65
Online exclusive
206.00 
Save 20.35 (10%)
-
Add to cart
Chemical Identifiers
CAS917-92-0
IUPAC Name3,3-dimethylbut-1-yne
Molecular FormulaC6H10
InChI KeyPPWNCLVNXGCGAF-UHFFFAOYSA-N
SMILESCC(C)(C)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3730 to 1.3750 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only