Thermo Scientific™

SIA (succinimidyl iodoacetate)

Catalog number: 22349
Have Questions?
Thermo Scientific™

SIA (succinimidyl iodoacetate)

Catalog number: 22349
Have Questions?
Catalog Number
22349
Unit Size
50 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2234950 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce SIA is a very short (1.5 angstrom) crosslinker for amine-to-sulfhydryl conjugation via N-hydroxysuccinimide (NHS) ester and iodoacetyl reactive groups.

Features of SIA:

Reactive groups: NHS ester and iodoacetyl
Reactive toward: amino and sulfhydryl groups
• Shortest sulfhydryl-reactive and amine-reactive heterobifunctional crosslinker available; compare to SIAB
• NHS ester reacts with primary amines at pH 7-9 to form a stable amide bond
• Iodoacetyl group reacts with sulfhydryl groups at pH > 7.5 to form stable thioether bond
• Non-cleavable
• Water-insoluble (dissolve first in DMF or DMSO)

Product References:

Crosslinker Application Guide -- search for recent literature references for this product
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine-Sulfhydryl
Cleavable
No
Description
SIA
Molecular Weight (g/mol)
283.02
PEGylated
No
Spacer Arm Length
1.5 Å
Cell Permeability
Yes
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
Haloacetyl, NHS Ester
Spacer
Short (<10 Å)
Form
Solid
Quantity
50 mg
Solubility
DMF, DMSO
Format
Standard
Water Soluble
No
Product Type
Crosslinker

Contents & Storage

Upon receipt store at 4°C protected from moisture.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text