Methyl 2-butynoate, 97%, Thermo Scientific Chemicals
Methyl 2-butynoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-butynoate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 223490250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS23326-27-4
IUPAC Namemethyl but-2-ynoate
Molecular FormulaC5H6O2
InChI KeyUJQCANQILFWSDJ-UHFFFAOYSA-N
SMILESCOC(=O)C#CC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4360 to 1.4380 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only