Nalgene™ Polypropylene, Wide-Mouth Carboy with Handle
Nalgene™ Polypropylene, Wide-Mouth Carboy with Handle
Thermo Scientific™

Nalgene™ Polypropylene, Wide-Mouth Carboy with Handle

For Use With: Store and transport solids, powders and liquids; Autoclavable; Closure Material: Polypropylene; Color: White
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
2235-002010 L
2235-0020PK10 L
2235-005020 L
2235-0050PK20 L
Catalog number 2235-0020
Price (EUR)
483,00
Case of 6
-
Add to cart
Capacity (Metric):
10 L
Recurring order eligible. Learn more »
Price (EUR)
483,00
Case of 6
Add to cart
Ask our AI about this Product

Store and transport solids, powders and liquids using Thermo Scientific™ Nalgene™ Wide-Mouth Autoclavable Polypropylene Carboys with Handles. These durable natural polypropylene (PP) carboys feature a convenient wide-mouth opening to promote ease of cleaning and dispensing. The wide-shoulder handles enable easy carrying and pouring, even with gloved hands.

  • Graduated to contain in 1 gal. and 5 L increments for ease of use
  • Wide-mouth opening can accommodate overhead mixers
  • Autoclavable
  • Leakproof† white polypropylene 100 mm screw closures

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity (English)2.6 gal.
Closure MaterialPolypropylene
Closure Size100-415 mm
DescriptionAutoclavable Wide-Mouth Carboy with Handles
Diameter (Metric) Outer250 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
GraduationsGraduated to contain in 5 L/1 gal. increments
Height (Metric) With Closure343 mm
IncludesHandles
No. per Case6/Cs.
No. per Pack1 Ea.
Volume (English) Brim3 gal.
Volume (Metric) Brim12 L
AutoclavableAutoclavable
Capacity (Metric)10 L
ColorWhite
MaterialPolypropylene
Mouth StyleWide
Product TypeCarboy with Handle
SterilityNon-sterile
Unit SizeCase of 6
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
1416168Certificate of ComplianceOct 29, 20242235-0050
1414871Certificate of ComplianceOct 10, 20242235-0050
1271371Certificate of ComplianceSep 10, 20242235-0020
1409588Certificate of ComplianceAug 16, 20242235-0050
1405626Certificate of ComplianceMay 10, 20242235-0020
5 results displayed, search above for a specific certificate

Frequently asked questions (FAQs)

Nalgene polypropylene carboys are safe to lift by the handles when full, as they are capable of supporting at least double their intended weight on a single handle.
To test the strength of the carboy handles, carboys are suspended through a single handle on the carboy body. 10 L, 20 L, and 50 L carboys are tested by filling the container with steel ballast pellets to a weight in excess of double the weight of the carboy filled to nominal capacity with water.

Share catalog number, name or link

1x1 image pixel for data collection