Thermo Scientific Chemicals

Antimony(III) sulfide, 98%, Thermo Scientific Chemicals

Catalog number: 223595000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Antimony(III) sulfide, 98%, Thermo Scientific Chemicals

Catalog number: 223595000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 223595000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1345-04-6
IUPAC Name
diantimony(3+) trisulfanediide
Molecular Formula
S3Sb2
InChI Key
NVWBARWTDVQPJD-UHFFFAOYSA-N
SMILES
[S--].[S--].[S--].[Sb+3].[Sb+3]
Appearance (Color)
Dark grey to black
Assay
>=97.5 % (Sb2S3)
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text