Thermo Scientific Chemicals

Manganese(II) chloride, 99+%, anhydrous, -80 mesh, Thermo Scientific Chemicals

Catalog number: 223610100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Manganese(II) chloride, 99+%, anhydrous, -80 mesh, Thermo Scientific Chemicals

Catalog number: 223610100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 223610100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7773-01-5
IUPAC Name
manganese(2+) dichloride
Molecular Formula
Cl2Mn
InChI Key
GLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Mn++]
Appearance (Color)
Pink
Titration Complexometric
>=99.0 %
Solubility
(10 % in dilute HCl) Clear solution
Particle size
-80 mesh
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text