Manganese(II) chloride, 99+%, anhydrous, -80 mesh, Thermo Scientific Chemicals
Manganese(II) chloride, 99+%, anhydrous, -80 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) chloride, 99+%, anhydrous, -80 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 223610500
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
Add to cart
Manganese(II) chloride, 99+%, anhydrous, -80 mesh, Thermo Scientific Chemicals
Catalog number223610500
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7773-01-5
IUPAC Namemanganese(2+) dichloride
Molecular FormulaCl2Mn
InChI KeyGLFNIEUTAYBVOC-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Mn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink
Titration Complexometric>=99.0 %
Solubility(10 % in dilute HCl) Clear solution
Particle size-80 mesh
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only