Tin(II) sulfate, 95%, Thermo Scientific Chemicals
Tin(II) sulfate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(II) sulfate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 223665000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7488-55-3
IUPAC Nameλ²-tin(2+) sulfate
Molecular FormulaO4SSn
InChI KeyOBBXFSIWZVFYJR-UHFFFAOYSA-L
SMILES[Sn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Titration Iodimetric>=94.0 %
Appearance (Form)Powder or crystals and/or lumps
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only