Tin(IV) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Tin(IV) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(IV) oxide, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 223681000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS18282-10-5
IUPAC Namestannanedione
Molecular FormulaO2Sn
InChI KeyXOLBLPGZBRYERU-UHFFFAOYSA-N
SMILESO=[Sn]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light grey
Total trace metal impurities=<0.1 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only