Thermo Scientific Chemicals

1-Iodo-2-methylpropane, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 223745000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

1-Iodo-2-methylpropane, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 223745000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 223745000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
513-38-2
IUPAC Name
1-iodo-2-methylpropane
Molecular Formula
C4H9I
InChI Key
BTUGGGLMQBJCBN-UHFFFAOYSA-N
SMILES
CC(C)CI
Infrared spectrum
Conforms
Stabilizer
copper
Appearance (Color)
Clear colorless to yellow to orange to brown
GC
>=96.0 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text