Thermo Scientific Chemicals

Methyl 4-formylbenzoate, 99%, Thermo Scientific Chemicals

Catalog number: 223750100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 4-formylbenzoate, 99%, Thermo Scientific Chemicals

Catalog number: 223750100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 223750100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1571-08-0
IUPAC Name
methyl 4-formylbenzoate
Molecular Formula
C9H8O3
InChI Key
FEIOASZZURHTHB-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(C=O)C=C1
Appearance (Color)
White to cream to light green
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder and/or chunks
GC
>=98.5 %
Melting point
59°C to 63°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text