Thermo Scientific Chemicals

Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals

Catalog number: 223760500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals

Catalog number: 223760500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 223760500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3087-36-3
IUPAC Name
titanium(4+) tetraethanolate
Molecular Formula
C8H20O4Ti
InChI Key
JMXKSZRRTHPKDL-UHFFFAOYSA-N
SMILES
[Ti+4].CC[O-].CC[O-].CC[O-].CC[O-]
Appearance (Color)
Clear colorless to yellow
Infrared spectrum
Conforms
Titration Complexometric
33 to 35 % (TiO2)
Appearance (Form)
Liquid
Refractive index
1.5030 to 1.5070 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text