Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals
Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number 223762500
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
Add to cart
Titanium(IV) ethoxide, 33-35% TiO2, Thermo Scientific Chemicals
Catalog number223762500
Price (USD)/ Each
159.65
Online exclusive
177.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS3087-36-3
IUPAC Nametitanium(4+) tetraethanolate
Molecular FormulaC8H20O4Ti
InChI KeyJMXKSZRRTHPKDL-UHFFFAOYSA-N
SMILES[Ti+4].CC[O-].CC[O-].CC[O-].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Titration Complexometric33 to 35 % (TiO2)
Appearance (Form)Liquid
Refractive index1.5030 to 1.5070 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only