Lithium tetrafluoroborate, 98%, pure, anhydrous, Thermo Scientific Chemicals
Lithium tetrafluoroborate, 98%, pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium tetrafluoroborate, 98%, pure, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Plastic bottle
Catalog number 223770500
Price (USD)/ Each
175.65
Online exclusive
195.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
175.65
Online exclusive
195.00 
Save 19.35 (10%)
Add to cart
Lithium tetrafluoroborate, 98%, pure, anhydrous, Thermo Scientific Chemicals
Catalog number223770500
Price (USD)/ Each
175.65
Online exclusive
195.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS14283-07-9
IUPAC Namelithium(1+) tetrafluoroboranuide
Molecular FormulaBF4Li
InChI KeyUFXJWFBILHTTET-UHFFFAOYSA-N
SMILES[Li+].F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)White to grey
Elemental Analysis>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only