Cadmium oxide, 99%, Pure, Thermo Scientific Chemicals
Cadmium oxide, 99%, Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium oxide, 99%, Pure, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 223790050
Price (USD)/ Each
271.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
319.00 
Save 47.35 (15%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
271.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
319.00 
Save 47.35 (15%)
Add to cart
Cadmium oxide, 99%, Pure, Thermo Scientific Chemicals
Catalog number223790050
Price (USD)/ Each
271.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
319.00 
Save 47.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1306-19-0
IUPAC Namecadmium(2+) oxidandiide
Molecular FormulaCdO
InChI KeyCFEAAQFZALKQPA-UHFFFAOYSA-N
SMILES[O--].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=98.5 %
Appearance (Form)Powder
Appearance (Color)Red-brown to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only