Thermo Scientific Chemicals

Potassium antimonyl tartrate sesquihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 223800025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Potassium antimonyl tartrate sesquihydrate, 99+%, Thermo Scientific Chemicals

Catalog number: 223800025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 223800025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28300-74-5
IUPAC Name
dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
Molecular Formula
C8H10K2O15Sb2
InChI Key
WBTCZEPSIIFINA-UHFFFAOYNA-J
SMILES
O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
Appearance (Color)
White
Infrared spectrum
Conforms
Loss on drying
=<2.7 % (105°C)
Titration Iodometric
>=99.0 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text