Chemical Identifiers
CAS28300-74-5
IUPAC Namedipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
Molecular FormulaC8H10K2O15Sb2
InChI KeyWBTCZEPSIIFINA-UHFFFAOYNA-J
SMILESO.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
View more