Potassium, 98%, chunks, in mineral oil, Thermo Scientific Chemicals
Potassium, 98%, chunks, in mineral oil, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium, 98%, chunks, in mineral oil, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 223860500
Price (USD)/ Each
141.65
Online exclusive
157.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
141.65
Online exclusive
157.00 
Save 15.35 (10%)
Add to cart
Potassium, 98%, chunks, in mineral oil, Thermo Scientific Chemicals
Catalog number223860500
Price (USD)/ Each
141.65
Online exclusive
157.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-09-7
IUPAC Namepotassium
Molecular FormulaK
InChI KeyZLMJMSJWJFRBEC-UHFFFAOYSA-N
SMILES[K]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Chunks
Appearance (Color)Grey
Total trace metal impurities=<2 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only