Chemical Identifiers
CAS14221-02-4
IUPAC Nametetrakis(triphenylphosphane) platinum
Molecular FormulaC72H60P4Pt
InChI KeySYKXNRFLNZUGAJ-UHFFFAOYSA-N
SMILES[Pt].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more