Bis(1,5-cyclooctadiene)nickel(0), Thermo Scientific Chemicals
Bis(1,5-cyclooctadiene)nickel(0), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(1,5-cyclooctadiene)nickel(0), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number 223970050
Price (USD)/ Each
238.65
Online exclusive
281.00 
Save 42.35 (15%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
238.65
Online exclusive
281.00 
Save 42.35 (15%)
Add to cart
Bis(1,5-cyclooctadiene)nickel(0), Thermo Scientific Chemicals
Catalog number223970050
Price (USD)/ Each
238.65
Online exclusive
281.00 
Save 42.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namebis((1Z,5Z)-cycloocta-1,5-diene) nickel
Molecular FormulaC16H24Ni
InChI KeyJRTIUDXYIUKIIE-KZUMESAESA-N
SMILES[Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Molecular Weight (g/mol)275.06
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Nickel (Ni)20.0 to 22.6 % (with dimethylglyoxime)
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only