Thermo Scientific Chemicals

Dirhenium decacarbonyl, sublimed, Thermo Scientific Chemicals

Catalog number: 223990050
Each
Thermo Scientific Chemicals

Dirhenium decacarbonyl, sublimed, Thermo Scientific Chemicals

Catalog number: 223990050
Each
Catalog number: 223990050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14285-68-8
IUPAC Name
{[1,1,1,1,2,2,2,2,2-nonakis(hydroxymethyl)dirhenium-1-yl]methylidyne}-λ³-oxidane
Molecular Formula
C10O10Re2
InChI Key
ZIZHEHXAMPQGEK-UHFFFAOYSA-N
SMILES
[O]#C[Re](C#[O])(C#[O])(C#[O])(C#[O])[Re](C#[O])(C#[O])(C#[O])(C#[O])C#[O]
Appearance (Form)
Crystals
Appearance (Color)
Yellow-green
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text