Triruthenium dodecacarbonyl, 99%, Thermo Scientific Chemicals
Triruthenium dodecacarbonyl, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triruthenium dodecacarbonyl, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 224000010
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
Add to cart
Triruthenium dodecacarbonyl, 99%, Thermo Scientific Chemicals
Catalog number224000010
Price (USD)/ Each
121.65
Online exclusive
135.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS15243-33-1
IUPAC Namecyclotriruthenium; dodecakis(methanidylidyneoxidanium)
Molecular FormulaC12O12Ru3
InChI KeyNQZFAUXPNWSLBI-UHFFFAOYSA-N
SMILES[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Ru]1[Ru][Ru]1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Elemental Analysis>=98.5 %
Appearance (Color)Orange
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only