Thermo Scientific Chemicals

Methyltriphenylphosphonium iodide, 97%, Thermo Scientific Chemicals

Catalog number: 224030250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Methyltriphenylphosphonium iodide, 97%, Thermo Scientific Chemicals

Catalog number: 224030250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 224030250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2065-66-9
IUPAC Name
methyltriphenylphosphanium iodide
Molecular Formula
C19H18IP
InChI Key
JNMIXMFEVJHFNY-UHFFFAOYSA-M
SMILES
[I-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Powder
Melting point
183°C to 188°C
Titration Argentometric
>=96.0 %
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text