Thermo Scientific Chemicals

(R)-(+)-1,1'-Bi-2-naphthol, 99%, Thermo Scientific Chemicals

Catalog number: 224300250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

(R)-(+)-1,1'-Bi-2-naphthol, 99%, Thermo Scientific Chemicals

Catalog number: 224300250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 224300250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18531-94-7
IUPAC Name
[1,1'-binaphthalene]-2,2'-diol
Molecular Formula
C20H14O2
InChI Key
PPTXVXKCQZKFBN-UHFFFAOYSA-N
SMILES
OC1=CC=C2C=CC=CC2=C1C1=C(O)C=CC2=CC=CC=C12
Appearance (Form)
Crystalline powder or crystals
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
HPLC
>=98.5 %
Melting point
205°C to 211°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text