Thermo Scientific Chemicals

(S)-(-)-1,1'-Bi-2-naphthol, 99%, Thermo Scientific Chemicals

Catalog number: 224310050
5 g, Each
Thermo Scientific Chemicals

(S)-(-)-1,1'-Bi-2-naphthol, 99%, Thermo Scientific Chemicals

Catalog number: 224310050
5 g, Each
Quantity
Catalog number: 224310050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18531-99-2
IUPAC Name
[1,1'-binaphthalene]-2,2'-diol
Molecular Formula
C20H14O2
InChI Key
PPTXVXKCQZKFBN-UHFFFAOYSA-N
SMILES
OC1=CC=C2C=CC=CC2=C1C1=C(O)C=CC2=CC=CC=C12
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Melting point
205°C to 211°C
HPLC
>=98.5 %
Appearance (Color)
White to light grey

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text