(S)-(+)-α-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
(S)-(+)-α-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(+)-α-Methoxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 224490010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS26164-26-1
IUPAC Name(2S)-2-methoxy-2-phenylacetate
Molecular FormulaC9H9O3
InChI KeyDIWVBIXQCNRCFE-QMMMGPOBSA-M
SMILESCO[C@H](C([O-])=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point63°C to 68°C
Titration with NaOH>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only