2,6-Di-tert-butyl-4-methylpyridine, 98%, Thermo Scientific Chemicals
2,6-Di-tert-butyl-4-methylpyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,6-Di-tert-butyl-4-methylpyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 224520250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name2,6-di-tert-butyl-4-methylpyridine
Molecular FormulaC14H23N
InChI KeyHVHZEKKZMFRULH-UHFFFAOYSA-N
SMILESCC1=CC(=NC(=C1)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol)205.35
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to orange to light brown
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4760 to 1.4780 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only