Trimethylsilyl isocyanate, 94%, Thermo Scientific Chemicals
Trimethylsilyl isocyanate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trimethylsilyl isocyanate, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 224730250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameisocyanatotrimethylsilane
Molecular FormulaC4H9NOSi
InChI KeyNIZHERJWXFHGGU-UHFFFAOYSA-N
SMILESC[Si](C)(C)N=C=O
Molecular Weight (g/mol)115.21
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3950 to 1.3970 (20°C, 589 nm)
GC>=94 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only