Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Conduct large-volume media and culture preparation with these Nalgene™ Round Polycarbonate Clearboy™ Carboys, particularly when visual inspection is required. This polycarbonate Clearboy is durable, lighter and safer than glass, autoclavable for sterile applications and ideal for refrigerated or frozen storage of aqueous solutions such as sterile water, aqueous reagents and media.
Autoclavable | Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | Clear |
Description | Round Transparent Clearboy |
Diameter (Metric) Outer | 253 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation |
Height (Metric) With Closure | 294 mm |
Material | Polycarbonate |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy |
Sterility | Non-sterile |
Volume (English) Brim | 3.3 gal. |
Volume (Metric) Brim | 12.5 L |
Unit Size | Case of 4 |
Autoclavable | Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | Clear |
Description | Round Transparent Clearboy |
Diameter (Metric) Outer | 253 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation |
Height (Metric) With Closure | 294 mm |
Material | Polycarbonate |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy |
Sterility | Non-sterile |
Volume (English) Brim | 3.3 gal. |
Volume (Metric) Brim | 12.5 L |
Unit Size | Each |
Autoclavable | Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | Clear |
Description | Round Transparent Clearboy |
Diameter (Metric) Outer | 287 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation |
Height (Metric) With Closure | 536 mm |
Material | Polycarbonate |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy |
Sterility | Non-sterile |
Volume (English) Brim | 6 gal. |
Volume (Metric) Brim | 24 L |
Unit Size | Case of 4 |
Autoclavable | Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Material | Polypropylene |
Closure Size | 83B |
Color | Clear |
Description | Round Transparent Clearboy |
Diameter (Metric) Outer | 287 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage, Transportation |
Height (Metric) With Closure | 536 mm |
Material | Polycarbonate |
No. per Case | 4/Cs. |
No. per Pack | 1 Ea. |
Product Type | Carboy |
Sterility | Non-sterile |
Volume (English) Brim | 6 gal. |
Volume (Metric) Brim | 24 L |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2251-0020 | Case of 4 | 10 L | 914.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2251-0020PK | Each | 10 L | 249.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2251-0050 | Case of 4 | 20 L | 1,304.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2251-0050PK | Each | 20 L | 352.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features:
Includes:
Ordering Information:
Additional closure options include Nalgene Quick Filling/Venting Closures (Cat. No. 2158), Nalgene Filling/Venting Closures (Cat. No. 2162) Nalgene Top Works™ Fluid Transfer Systems (Cat. No. 2135) and Nalgene Validation Bottles (Cat. No. DS2127). Additional accessories include Nalgene Barbed Bulk Head Fittings (Cat. No. 6149).
Note:
Completely disengage closure threads and disassemble spigot before autoclaving
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.