3-Aminosalicylic acid, 97%, Thermo Scientific Chemicals
3-Aminosalicylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminosalicylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
50 mg
250 mg
Packaging:
Glass bottle
Catalog number 225260010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS570-23-0
IUPAC Name3-amino-2-hydroxybenzoic acid
Molecular FormulaC7H7NO3
InChI KeyIQGMRVWUTCYCST-UHFFFAOYSA-N
SMILESNC1=CC=CC(C(O)=O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Form)Powder
Appearance (Color)Beige to grey to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only