Thermo Scientific Chemicals

3-Aminosalicylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 225260500
50 mg, Glass bottle, Each
Thermo Scientific Chemicals

3-Aminosalicylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 225260500
50 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 225260500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
570-23-0
IUPAC Name
3-amino-2-hydroxybenzoic acid
Molecular Formula
C7H7NO3
InChI Key
IQGMRVWUTCYCST-UHFFFAOYSA-N
SMILES
NC1=CC=CC(C(O)=O)=C1O
HPLC
>=96.0 %
Appearance (Form)
Powder
Appearance (Color)
Beige to grey to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text