Thermo Scientific Chemicals

Calcium sulfate dihydrate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 225270250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium sulfate dihydrate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 225270250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 225270250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10101-41-4
IUPAC Name
calcium dihydrate sulfate
Molecular Formula
CaH4O6S
InChI Key
PASHVRUKOFIRIK-UHFFFAOYSA-L
SMILES
O.O.[Ca++].[O-]S([O-])(=O)=O
Titration Complexometric
98.0 to 102.0 %
Insoluble matter
=<0.05 % (in dilute HCl)
Appearance (Color)
White
Heavy metals (as Pb)
=<20 ppm
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text