Calcium sulfate dihydrate, 98+%, for analysis
Calcium sulfate dihydrate, 98+%, for analysis
Calcium sulfate dihydrate, 98+%, for analysis
Calcium sulfate dihydrate, 98+%, for analysis
Thermo Scientific Chemicals

Calcium sulfate dihydrate, 98+%, for analysis

CAS: 10101-41-4 | CaH4O6S | 172.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 225275000
Price (USD)
73.65
Online Exclusive
81.50
Save 7.85 (10%)
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
73.65
Online Exclusive
81.50
Save 7.85 (10%)
Each
Add to cart
Chemical Identifiers
CAS10101-41-4
IUPAC Namecalcium dihydrate sulfate
Molecular FormulaCaH4O6S
InChI KeyPASHVRUKOFIRIK-UHFFFAOYSA-L
SMILESO.O.[Ca++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric98.0 to 102.0 %
Insoluble matter=<0.05 % (in dilute HCl)
Appearance (Form)Powder
Appearance (Color)White
Heavy metals (as Pb)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link