Thermo Scientific Chemicals

N,N-Dimethyloctylamine, 97%, Thermo Scientific Chemicals

Catalog number: 225290250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Dimethyloctylamine, 97%, Thermo Scientific Chemicals

Catalog number: 225290250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 225290250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7378-99-6
IUPAC Name
dimethyl(octyl)amine
Molecular Formula
C10H23N
InChI Key
UQKAOOAFEFCDGT-UHFFFAOYSA-N
SMILES
CCCCCCCCN(C)C
Color scale
=<30 APHA
Titration with HClO4
>=96.0 %
Appearance (Form)
Clear to slightly hazy liquid
Infrared spectrum
Conforms
Refractive index
1.4233 to 1.4253 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text