Thermo Scientific Chemicals

Chromium, 99+%, Thermo Scientific Chemicals

Catalog number: 225491000
100 g, Each
Thermo Scientific Chemicals

Chromium, 99+%, Thermo Scientific Chemicals

Catalog number: 225491000
100 g, Each
Quantity
Catalog number: 225491000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-47-3
IUPAC Name
chromium
Molecular Formula
Cr
InChI Key
VYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES
[Cr]
Titration with Na2S2O3
>=99.0 %
Appearance (Color)
Grey
Appearance (Form)
Metallic powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text