3,4-Dihydroxyphenylacetic acid, 98%, Thermo Scientific Chemicals
3,4-Dihydroxyphenylacetic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dihydroxyphenylacetic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 225600050
Price (USD)/ Each
109.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
109.00
Add to cart
3,4-Dihydroxyphenylacetic acid, 98%, Thermo Scientific Chemicals
Catalog number225600050
Price (USD)/ Each
109.00
-
Add to cart
Chemical Identifiers
IUPAC Name2-(3,4-dihydroxyphenyl)acetic acid
Molecular FormulaC8H8O4
InChI KeyCFFZDZCDUFSOFZ-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=C(O)C(O)=C1
Molecular Weight (g/mol)168.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point127°C to 132°C
HPLC>=97.5 %
Appearance (Color)Beige to light brown to purple-grey
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only