Thermo Scientific Chemicals

D(+)-Melezitose monohydrate, 99+%

Catalog number: 225640050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

D(+)-Melezitose monohydrate, 99+%

Catalog number: 225640050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 225640050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10030-67-8
IUPAC Name
2-{[4-hydroxy-2,5-bis(hydroxymethyl)-2-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxolan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol dihydrate
Molecular Formula
C18H36O18
InChI Key
LNVIPYYEBMNJIL-UHFFFAOYNA-N
SMILES
O.O.OCC1OC(CO)(OC2OC(CO)C(O)C(O)C2O)C(OC2OC(CO)C(O)C(O)C2O)C1O
Appearance (Form)
Crystalline powder
HPLC
>=99.0 %
Loss on drying
=<4.5 % (110°C, 1 mmHg)
Appearance (Color)
White
Specific optical rotation
+86° to +90° (20°C, 589 nm) (c=4, water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Sap-feeding insects such as ants and aphids produce melezitose to maintain osmotic balance

• Melezitose is produced by some plants as an insect attractant and acts as a metabolite

Applications

• Melezitose can be hydrolyzed to glucose and turanose through alpha-glucosidase activity and can be used for the study of enzyme activity

• Melezitose can be used to distinguish certain strains of bacteria; characterization of phenotype based on the presence or absence of certain sugars assists identification

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text