Thermo Scientific Chemicals

Diphenyl phosphate, 99%, Thermo Scientific Chemicals

Catalog number: 225690250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Diphenyl phosphate, 99%, Thermo Scientific Chemicals

Catalog number: 225690250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 225690250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
838-85-7
IUPAC Name
diphenoxyphosphinic acid
Molecular Formula
C12H11O4P
InChI Key
ASMQGLCHMVWBQR-UHFFFAOYSA-N
SMILES
OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
Appearance (Color)
White
Melting point
62°C to 70°C
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Titration with NaOH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text