Thermo Scientific Chemicals

Adenosine 3',5'-cyclic monophosphate, 99+%

Catalog number: 225800050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Adenosine 3',5'-cyclic monophosphate, 99+%

Catalog number: 225800050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 225800050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
60-92-4
IUPAC Name
(4aR,6R,7R,7aS)-6-(6-amino-9H-purin-9-yl)-2,7-dihydroxy-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-2-one
Molecular Formula
C10H12N5O6P
InChI Key
IVOMOUWHDPKRLL-YPLCUDRINA-N
SMILES
NC1=C2N=CN([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1
Ratio of Absorbances
lambda max. 260 nm, pH 7
Ratio of Absorbances
0.13 to 0.19 (A280/A260)
Ratio of Absorbances
0.77 to 0.83 (A250/A260)
Appearance (Form)
Crystalline powder
Water
=<7 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text