S-Carboxymethyl-L-cysteine, 98%, Thermo Scientific Chemicals
S-Carboxymethyl-L-cysteine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

S-Carboxymethyl-L-cysteine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 225810050
Price (USD)/ Each
23.20
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
23.20
Add to cart
S-Carboxymethyl-L-cysteine, 98%, Thermo Scientific Chemicals
Catalog number225810050
Price (USD)/ Each
23.20
-
Add to cart
Chemical Identifiers
CAS638-23-3
IUPAC Name(2R)-2-amino-3-[(carboxymethyl)sulfanyl]propanoic acid
Molecular FormulaC5H9NO4S
InChI KeyGBFLZEXEOZUWRN-VKHMYHEASA-N
SMILESN[C@@H](CSCC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
Loss on drying=<0.5 % (105°C, 3 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only