Thermo Scientific Chemicals

S-Carboxymethyl-L-cysteine, 98%, Thermo Scientific Chemicals

Catalog number: 225811000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

S-Carboxymethyl-L-cysteine, 98%, Thermo Scientific Chemicals

Catalog number: 225811000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 225811000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
638-23-3
IUPAC Name
(2R)-2-amino-3-[(carboxymethyl)sulfanyl]propanoic acid
Molecular Formula
C5H9NO4S
InChI Key
GBFLZEXEOZUWRN-VKHMYHEASA-N
SMILES
N[C@@H](CSCC(O)=O)C(O)=O
Appearance (Color)
White to almost white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with HClO4
97.5 to 102.5 %
Loss on drying
=<0.5 % (105°C, 3 h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text