L(-)-Fucose, 97%
Thermo Scientific Chemicals

L(-)-Fucose, 97%

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 225880010
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.30 
Save 8.65 (14%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.30 
Save 8.65 (14%)
Add to cart
L(-)-Fucose, 97%
Catalog number225880010
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.30 
Save 8.65 (14%)
-
Add to cart
Chemical Identifiers
CAS2438-80-4
IUPAC Name(3S,4R,5S,6S)-6-methyloxane-2,3,4,5-tetrol
Molecular FormulaC6H12O5
InChI KeySHZGCJCMOBCMKK-DHVFOXMCSA-N
SMILESC[C@@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-72° to -78° (20°C, 589 nm) (c=4, H2O, NH3)
Appearance (Color)White to off-white
Infrared spectrumConforms
Water=<1 % (K.F.)
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only